Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF144A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014746.4 | NP_055561.2 | 6 | 859 | ||
XM_005246200.3 | XP_005246257.1 | 6 | 2050 | |||
XM_005246202.4 | XP_005246259.1 | 5 | 690 | |||
XM_017005396.1 | XP_016860885.1 | 6 | 2026 | |||
XM_017005397.1 | XP_016860886.1 | 6 | 684 | |||
XM_017005398.1 | XP_016860887.1 | 6 | 797 | |||
XM_017005399.1 | XP_016860888.1 | 6 | 797 | |||
XM_017005400.1 | XP_016860889.1 | 6 | 797 | |||
XM_017005401.1 | XP_016860890.1 | 6 | 797 | |||
XM_017005402.1 | XP_016860891.1 | 6 | 797 | |||
XM_017005403.1 | XP_016860892.1 | 6 | 795 | |||
XM_017005404.1 | XP_016860893.1 | 5 | 667 | |||
GenBank mRNA | AK293854.1 | - | 6 | 796 | ||
BC050373.1 | - | 6 | 799 | 5722878 | ||
D79983.1 | - | 6 | 765 |