Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014765.2 | NP_055580.1 | 5 | 1167 | ||
GenBank mRNA | BC100286.1 | - | 5 | 1049 | 6157036 | |
D13641.1 | - | 5 | 1043 |