Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039459.2 | NP_001034548.1 | 5 | 629 | ||
NM_001206480.2 | NP_001193409.1 | 20 | 2408 | |||
NM_001206482.1 | NP_001193411.1 | 20 | 2578 | |||
NM_014800.10 | NP_055615.8 | 20 | 2219 | |||
NM_130442.3 | NP_569709.1 | 5 | 775 | |||
NR_038120.1 | 4 | 662 | ||||
XM_005249919.2 | XP_005249976.1 | 21 | 2772 | |||
XM_006715805.1 | XP_006715868.1 | 20 | 2064 | |||
XM_011515654.1 | XP_011513956.1 | 21 | 3457 | |||
XM_017012838.1 | XP_016868327.1 | 21 | 3642 | |||
XM_017012839.1 | XP_016868328.1 | 20 | 2006 | |||
GenBank mRNA | AF398885.1 | - | 19 | 1870 | ||
AK126565.1 | - | 8 | 1259 | |||
AK291784.1 | - | 20 | 2578 | |||
AK296253.1 | - | 17 | 1739 | |||
AK301151.1 | - | 4 | 537 | |||
AK314349.1 | - | 20 | 2008 | |||
AL136787.1 | - | 20 | 2120 | |||
BC003051.1 | - | 5 | 625 | |||
BC064635.1 | - | 5 | 577 | |||
BC077074.1 | - | 5 | 626 | |||
BC114341.2 | - | 20 | 1957 | 40035461 | ||
D87457.1 | - | 5 | 601 |