Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC24D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318066.1 | NP_001304995.1 | 23 | 3575 | ||
NM_014822.3 | NP_055637.2 | 23 | 3572 | |||
XM_005263379.2 | XP_005263436.1 | 23 | 3449 | |||
XM_017008875.1 | XP_016864364.1 | 18 | 2730 | |||
GenBank mRNA | AB018298.1 | - | 23 | 3475 | ||
AK001390.1 | - | 20 | 2943 | |||
AK125962.1 | - | 18 | 3407 | |||
AK291765.1 | - | 23 | 3472 | |||
AK309960.1 | - | 16 | 2311 | |||
BC035761.1 | - | 23 | 3497 | 5554774 |