Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM65B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286445.1 | NP_001273374.1 | 11 | 1063 | ||
NM_001286446.1 | NP_001273375.1 | 11 | 1166 | |||
NM_001286447.1 | NP_001273376.1 | 11 | 1263 | |||
NM_014722.3 | NP_055537.2 | 11 | 1201 | |||
NM_015864.3 | NP_056948.2 | 11 | 1263 | |||
XM_006715275.2 | XP_006715338.1 | 11 | 1280 | |||
XM_006715277.1 | XP_006715340.1 | 11 | 1201 | |||
XM_006715279.2 | XP_006715342.1 | 11 | 1258 | |||
XM_006715281.3 | XP_006715344.1 | 11 | 1280 | |||
XM_011515007.1 | XP_011513309.1 | 12 | 1789 | |||
XM_011515008.2 | XP_011513310.1 | 12 | 1120 | |||
XM_011515009.1 | XP_011513311.1 | 11 | 992 | |||
XM_011515010.2 | XP_011513312.1 | 11 | 1033 | |||
XM_011515012.1 | XP_011513314.1 | 11 | 1280 | |||
XM_017011523.1 | XP_016867012.1 | 11 | 955 | |||
XM_017011524.1 | XP_016867013.1 | 11 | 1209 | |||
XM_017011525.1 | XP_016867014.1 | 11 | 1201 | |||
XM_017011526.1 | XP_016867015.1 | 11 | 1201 | |||
XM_017011527.1 | XP_016867016.1 | 11 | 1033 | |||
GenBank mRNA | AB002384.1 | - | 11 | 999 | ||
AK299047.1 | - | 11 | 971 | |||
AK300138.1 | - | 11 | 1211 | |||
AK300950.1 | - | 11 | 1016 | |||
BC001232.1 | - | 11 | 1253 | 3453890 | ||
U49187.1 | - | 11 | 1124 |