Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003889.3 | NP_003880.3 | 8 | 2948 | ||
NM_022002.2 | NP_071285.1 | 8 | 1274 | |||
NM_033013.2 | NP_148934.1 | 8 | 2837 | |||
GenBank mRNA | AB307701.1 | - | 7 | 1109 | ||
AF061056.1 | - | 8 | 1412 | |||
AF084644.1 | - | 8 | 1285 | |||
AF084645.1 | - | 8 | 1388 | |||
AJ009936.1 | - | 9 | 2948 | |||
AJ009937.1 | - | 9 | 2837 | |||
AK122990.1 | - | 5 | 1241 | |||
AY091855.1 | - | 8 | 1691 | |||
BC017304.2 | - | 8 | 1345 | 5087401 | ||
HQ692837.1 | - | 8 | 1226 | |||
HQ709177.1 | - | 8 | 1217 |