Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAZ1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032408.3 | NP_115784.1 | 3 | 594 | ||
XM_017012773.1 | XP_016868262.1 | 3 | 607 | |||
GenBank mRNA | AB032253.1 | - | 3 | 587 | ||
AF072810.1 | - | 3 | 594 | |||
AF084479.1 | - | 3 | 526 | |||
BC065029.1 | - | 4 | 296 | |||
BC136520.1 | - | 3 | 427 | 9020510 |