Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GIT2 SIRNA Assays ›
Gene Symbol:
GIT2
Gene Name:
GIT ArfGAP 2
Gene Aliases:
CAT-2, CAT2, PKL
Chromosome Location:
Chr.12: 109929792 - 109996389 on Build GRCh38
UniGene ID:
Hs.434996
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138618
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GIT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135213.1
NP_001128685.1
3
370
NM_001135214.1
NP_001128686.1
3
370
NM_014776.3
NP_055591.2
3
370
NM_057169.3
NP_476510.1
3
370
NM_057170.3
NP_476511.1
3
370
NM_139201.2
NP_631940.2
3
370
XM_005253997.3
XP_005254054.1
3
365
XM_005254000.3
XP_005254057.1
3
365
XM_006719707.3
XP_006719770.1
3
365
XM_006719708.3
XP_006719771.1
3
365
XM_006719709.3
XP_006719772.1
3
365
XM_006719712.3
XP_006719775.1
3
365
XM_006719713.3
XP_006719776.1
3
362
XM_006719714.3
XP_006719777.1
3
365
XM_017020258.1
XP_016875747.1
3
365
XM_017020259.1
XP_016875748.1
3
362
XM_017020260.1
XP_016875749.1
3
362
XM_017020261.1
XP_016875750.1
3
360
GenBank mRNA
AB451294.1
-
3
205
AB451428.1
-
3
205
AF124491.1
-
3
205
AK222736.1
-
3
277
AK303198.1
-
2
71
AK304240.1
-
3
354
AK308304.1
-
3
285
BC001379.2
-
3
270
3051069
BC014223.2
-
2
121
BP871813.1
-
3
370
BT007312.1
-
3
205
CR533568.1
-
3
205
D63482.2
-
3
276
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of GTPase activity
behavioral response to pain
GTPase activator activity
protein binding
protein complex binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0