Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261464.1 | NP_001248393.1 | 21 | 2626 | ||
NM_001261465.1 | NP_001248394.1 | 21 | 2598 | |||
NM_003815.4 | NP_003806.3 | 21 | 2596 | |||
NM_207191.2 | NP_997074.1 | 20 | 2526 | |||
NM_207194.2 | NP_997077.1 | 22 | 2671 | |||
NM_207195.2 | NP_997078.1 | 22 | 2668 | |||
NM_207196.2 | NP_997079.1 | 23 | 2740 | |||
NM_207197.2 | NP_997080.1 | 23 | 2743 | |||
NR_048577.1 | 23 | 2882 | ||||
NR_048578.1 | 23 | 2885 | ||||
NR_048579.1 | 22 | 2810 | ||||
GenBank mRNA | AB209157.1 | - | 22 | 4271 | ||
AK000667.1 | - | 13 | 1292 | |||
AK075498.1 | - | 13 | 1670 | |||
AK296925.1 | - | 21 | 2547 | |||
AY560593.1 | - | 20 | 2512 | |||
AY560594.1 | - | 21 | 2582 | |||
AY560595.1 | - | 21 | 2584 | |||
AY560596.1 | - | 22 | 2657 | |||
AY560597.1 | - | 22 | 2654 | |||
AY560598.1 | - | 23 | 2726 | |||
AY560599.1 | - | 23 | 2729 | |||
AY560600.1 | - | 23 | 2868 | |||
AY560601.1 | - | 23 | 2871 | |||
AY576417.1 | - | 22 | 2796 | |||
BC014566.2 | - | 21 | 2562 | 2958341 | ||
U41767.1 | - | 21 | 2488 | |||
U46005.1 | - | 21 | 2481 |