Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000358.2 | NP_000349.1 | 5 | 651 | ||
GenBank mRNA | AB209598.1 | - | 1 | 1137 | ||
AK093916.1 | - | 5 | 506 | |||
AK094055.1 | - | 6 | 582 | |||
AK094581.1 | - | 5 | 595 | |||
AK222833.1 | - | 5 | 559 | |||
AK303643.1 | - | 5 | 883 | |||
BC000097.2 | - | 5 | 544 | 3506051 | ||
BC004972.1 | - | 5 | 539 | 2958878 | ||
BT009820.1 | - | 5 | 490 | |||
KU178357.1 | - | 5 | 490 | |||
KU178358.1 | - | 5 | 490 | |||
M77349.1 | - | 5 | 537 |