Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000358.2 | NP_000349.1 | 17 | 2215 | ||
GenBank mRNA | AK093916.1 | - | 17 | 2070 | ||
AK094055.1 | - | 18 | 2146 | |||
AK094581.1 | - | 17 | 2159 | |||
AK222833.1 | - | 17 | 2123 | |||
AK298603.1 | - | 12 | 1435 | |||
AK303643.1 | - | 17 | 2447 | |||
BC000097.2 | - | 17 | 2108 | 3506051 | ||
BC004972.1 | - | 17 | 2103 | 2958878 | ||
M77349.1 | - | 17 | 2101 |