Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other THRB SIRNA Assays ›
Gene Symbol:
THRB
Gene Name:
thyroid hormone receptor beta
Gene Aliases:
C-ERBA-2, C-ERBA-BETA, ERBA2, GRTH, NR1A2, PRTH, THR1, THRB1, THRB2
Chromosome Location:
Chr.3: 24117153 - 24495281 on Build GRCh38
UniGene ID:
Hs.187861
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138738
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: THRB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000461.4
NP_000452.2
4
396
NM_001128176.2
NP_001121648.1
5
507
NM_001128177.1
NP_001121649.1
5
533
NM_001252634.1
NP_001239563.1
6
631
XM_005265421.4
XP_005265478.1
5
604
XM_005265423.4
XP_005265480.1
4
439
XM_005265424.3
XP_005265481.1
4
374
XM_006713317.3
XP_006713380.1
5
747
XM_006713318.3
XP_006713381.1
5
699
XM_011534046.2
XP_011532348.1
5
2647
XM_011534047.2
XP_011532349.1
6
884
XM_011534048.2
XP_011532350.1
5
463
XM_011534049.2
XP_011532351.1
4
319
XM_011534050.2
XP_011532352.1
5
692
XM_011534051.2
XP_011532353.1
4
282
XM_011534052.2
XP_011532354.1
5
419
XM_011534053.2
XP_011532355.1
4
540
XM_017007108.1
XP_016862597.1
6
817
XM_017007109.1
XP_016862598.1
7
975
XM_017007110.1
XP_016862599.1
6
782
XM_017007111.1
XP_016862600.1
5
428
XM_017007112.1
XP_016862601.1
4
398
XM_017007113.1
XP_016862602.1
3
537
XM_017007114.1
XP_016862603.1
3
502
XM_017007115.1
XP_016862604.1
3
376
GenBank mRNA
AB307687.1
-
1
96
AK096628.1
-
6
620
AY286465.1
-
4
322
AY286466.1
-
4
340
AY286467.1
-
4
306
AY286468.1
-
5
425
AY286469.1
-
5
416
AY286470.1
-
5
450
AY286471.1
-
6
499
BC106929.1
-
3
221
40033198
BC106930.1
-
4
367
40033200
BP313943.1
-
4
385
BQ879477.1
-
4
518
DA218319.1
-
4
336
GQ919288.1
-
4
259
HQ692825.1
-
2
111
M26747.1
-
4
396
X04707.1
-
4
396
Back To Top
More Information
Panther Classification:
Molecular Function -
C4 zinc finger nuclear receptor
DNA-binding transcription factor
zinc finger transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
sensory perception of sound
negative regulation of female receptivity
regulation of heart contraction
female courtship behavior
organ morphogenesis
intracellular receptor signaling pathway
steroid hormone mediated signaling pathway
positive regulation of transcription from RNA polymerase II promoter
Type I pneumocyte differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
transcription corepressor activity
thyroid hormone receptor activity
protein binding
zinc ion binding
enzyme binding
chromatin DNA binding
sequence-specific DNA binding
thyroid hormone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-dvnl5:80/10.239.162.83:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0