Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSC2 SIRNA Assays ›
Gene Symbol:
TSC2
Gene Name:
tuberous sclerosis 2
Gene Aliases:
LAM, PPP1R160, TSC4
Chromosome Location:
Chr.16: 2047804 - 2088720 on Build GRCh38
UniGene ID:
Hs.90303
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138743
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000548.4
NP_000539.2
35
4744
NM_001077183.2
NP_001070651.1
33
4543
NM_001114382.2
NP_001107854.1
34
4675
NM_001318827.1
NP_001305756.1
32
4435
NM_001318829.1
NP_001305758.1
32
4379
NM_001318831.1
NP_001305760.1
31
4238
NM_001318832.1
NP_001305761.1
33
4444
XM_005255529.4
XP_005255586.2
34
4502
XM_005255531.4
XP_005255588.2
33
4433
XM_011522636.2
XP_011520938.1
35
4685
XM_011522637.2
XP_011520939.1
35
4682
XM_011522638.2
XP_011520940.2
34
4778
XM_011522639.2
XP_011520941.1
34
4556
XM_011522640.2
XP_011520942.1
34
4553
XM_017023615.1
XP_016879104.1
35
4628
XM_017023616.1
XP_016879105.1
34
4499
XM_017023617.1
XP_016879106.1
32
4526
XM_017023618.1
XP_016879107.1
36
4772
GenBank mRNA
AB210000.1
-
33
4449
AK094152.1
-
3
1659
AK125096.1
-
8
2302
AK295672.1
-
32
4266
AK295728.1
-
33
4444
AK299343.1
-
32
4316
BC025364.1
-
8
1463
BC046929.1
-
8
1463
BC150300.1
-
34
4550
8860381
BX647816.1
-
31
4101
KJ534954.1
-
32
4346
KJ534955.1
-
32
4343
KJ534958.1
-
32
4343
KJ535039.1
-
24
2769
KJ535091.1
-
33
4415
KJ535094.1
-
19
2730
X75621.1
-
34
4562
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
negative regulation of protein kinase activity
protein import into nucleus
endocytosis
cell cycle arrest
heart development
protein localization
negative regulation of cell proliferation
negative regulation of phosphatidylinositol 3-kinase signaling
viral process
vesicle-mediated transport
regulation of endocytosis
negative regulation of Wnt signaling pathway
negative regulation of TOR signaling
protein kinase B signaling
positive regulation of GTPase activity
regulation of insulin receptor signaling pathway
negative regulation of insulin receptor signaling pathway
insulin-like growth factor receptor signaling pathway
positive chemotaxis
regulation of small GTPase mediated signal transduction
regulation of cell cycle
negative regulation of protein kinase B signaling
GTPase activator activity
protein binding
phosphatase binding
small GTPase binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0