Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001057.2 | NP_001048.2 | 1 | 749 | ||
GenBank mRNA | AK291996.1 | - | 1 | 749 | ||
AK300649.1 | - | 1 | 643 | |||
AY322545.1 | - | 1 | 154 | |||
BC096842.1 | - | 1 | 160 | 6971906 | ||
M57414.1 | - | 1 | 154 |
Set Membership: |
![]() |