Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001057.2 | NP_001048.2 | 5 | 1744 | ||
GenBank mRNA | AK096906.1 | - | 3 | 769 | ||
AK291996.1 | - | 5 | 1744 | |||
AK300649.1 | - | 5 | 1542 | |||
AY322545.1 | - | 5 | 1149 | |||
BC096842.1 | - | 5 | 1155 | 6971906 | ||
FJ200479.1 | - | 6 | 1041 | |||
M57414.1 | - | 5 | 1149 |
Set Membership: |
![]() |