Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324282.1 | NP_001311211.1 | 4 | 454 | ||
NM_001324283.1 | NP_001311212.1 | 4 | 465 | |||
NM_001324284.1 | NP_001311213.1 | 6 | 654 | |||
NM_001324285.1 | NP_001311214.1 | 6 | 662 | |||
NM_001324286.1 | NP_001311215.1 | 2 | 293 | |||
NM_001324287.1 | NP_001311216.1 | 3 | 378 | |||
NM_001324288.1 | NP_001311217.1 | 5 | 550 | |||
NM_001324289.1 | NP_001311218.1 | 4 | 465 | |||
NM_003473.3 | NP_003464.1 | 5 | 550 | |||
XM_011519695.2 | XP_011517997.1 | 5 | 526 | |||
GenBank mRNA | AK303083.1 | - | 3 | 238 | ||
AK314405.1 | - | 5 | 434 | |||
AU118708.1 | - | 5 | 551 | |||
BC030586.2 | - | 5 | 493 | 4820857 | ||
U43899.1 | - | 5 | 377 |