Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135774.1 | NP_001129246.1 | 14 | 2729 | ||
NM_003490.3 | NP_003481.3 | 13 | 2629 | |||
NM_133633.2 | NP_598344.2 | 12 | 2337 | |||
XM_011530405.2 | XP_011528707.1 | 14 | 3265 | |||
XM_011530406.2 | XP_011528708.1 | 14 | 2731 | |||
XM_011530407.2 | XP_011528709.1 | 14 | 3138 | |||
XM_011530408.1 | XP_011528710.1 | 14 | 2729 | |||
XM_011530410.2 | XP_011528712.1 | 12 | 2933 | |||
XM_017028960.1 | XP_016884449.1 | 15 | 3663 | |||
XM_017028961.1 | XP_016884450.1 | 14 | 3055 | |||
XM_017028962.1 | XP_016884451.1 | 14 | 2990 | |||
XM_017028963.1 | XP_016884452.1 | 14 | 2654 | |||
XM_017028964.1 | XP_016884453.1 | 14 | 2726 | |||
XM_017028965.1 | XP_016884454.1 | 8 | 2500 |