Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DYSF SIRNA Assays ›
Gene Symbol:
DYSF
Gene Name:
dysferlin
Gene Aliases:
FER1L1, LGMD2B, MMD1
Chromosome Location:
Chr.2: 71453155 - 71686763 on Build GRCh38
UniGene ID:
Hs.252180
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139106
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DYSF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130455.1
NP_001123927.1
15
1531
NM_001130976.1
NP_001124448.1
15
1763
NM_001130977.1
NP_001124449.1
15
1763
NM_001130978.1
NP_001124450.1
15
1763
NM_001130979.1
NP_001124451.1
16
1856
NM_001130980.1
NP_001124452.1
16
1856
NM_001130981.1
NP_001124453.1
16
1856
NM_001130982.1
NP_001124454.1
16
1624
NM_001130983.1
NP_001124455.1
15
1531
NM_001130984.1
NP_001124456.1
15
1531
NM_001130985.1
NP_001124457.1
16
1624
NM_001130986.1
NP_001124458.1
15
1531
NM_001130987.1
NP_001124459.1
16
1624
NM_003494.3
NP_003485.1
15
1763
XM_005264584.4
XP_005264641.1
16
1639
XM_005264585.4
XP_005264642.1
16
2324
GenBank mRNA
AF075575.1
-
15
1760
AK074104.1
-
2
75
BC171803.1
-
15
1736
DQ267935.1
-
15
1531
EU515155.1
-
16
1757
EU515156.1
-
15
1664
EU515157.1
-
15
1664
EU515158.1
-
16
1757
EU515159.1
-
15
1664
EU515160.1
-
16
1757
EU515161.1
-
16
1624
EU515162.1
-
15
1531
EU515163.1
-
15
1531
EU515164.1
-
16
1624
EU515165.1
-
15
1531
EU515166.1
-
16
1624
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
plasma membrane repair
vesicle fusion
muscle contraction
calcium ion binding
protein binding
phospholipid binding
calcium-dependent phospholipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-kprhh:80/10.239.160.57:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline