Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261408.1 | NP_001248337.1 | 9 | 1575 | ||
NM_001261409.1 | NP_001248338.1 | 7 | 1561 | |||
NM_001261410.1 | NP_001248339.1 | 5 | 1228 | |||
NM_001308251.1 | NP_001295180.1 | 3 | 548 | |||
NM_003567.3 | NP_003558.1 | 7 | 1538 | |||
XM_011542251.2 | XP_011540553.1 | 6 | 1177 | |||
XM_017002480.1 | XP_016857969.1 | 9 | 1951 | |||
GenBank mRNA | AF124250.1 | - | 7 | 1530 | ||
AK027785.1 | - | 3 | 796 | |||
AK055442.1 | - | 7 | 1561 | |||
AK301570.1 | - | 3 | 521 | |||
AK310970.1 | - | 3 | 548 | |||
AL833121.1 | - | 5 | 1181 | |||
AY358996.1 | - | 3 | 468 | |||
BC028477.1 | - | 9 | 1604 | |||
BC039895.1 | - | 7 | 1431 | 6060541 | ||
DA745974.1 | - | 3 | 511 | |||
U92715.1 | - | 7 | 1346 |