Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DUSP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003584.2 | NP_003575.2 | 9 | 1177 | ||
GenBank mRNA | AF023917.1 | - | 9 | 1118 | ||
BC000346.2 | - | 9 | 1095 | 2960939 |