Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303040.1 | NP_001289969.1 | 27 | 3750 | ||
NM_001303041.1 | NP_001289970.1 | 26 | 3638 | |||
NM_003637.4 | NP_003628.2 | 30 | 4067 | |||
XM_005277436.3 | XP_005277493.1 | 29 | 3818 | |||
XM_011510083.2 | XP_011508385.1 | 26 | 3498 | |||
XM_011510084.1 | XP_011508386.1 | 19 | 2495 | |||
XM_017002622.1 | XP_016858111.1 | 28 | 4024 | |||
XM_017002623.1 | XP_016858112.1 | 28 | 4021 | |||
XM_017002624.1 | XP_016858113.1 | 27 | 3910 | |||
XM_017002625.1 | XP_016858114.1 | 27 | 3910 | |||
XM_017002626.1 | XP_016858115.1 | 26 | 3781 | |||
XM_017002628.1 | XP_016858117.1 | 24 | 3474 | |||
GenBank mRNA | AF112345.1 | - | 30 | 3968 |