Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320973.1 | NP_001307902.1 | 4 | 1141 | ||
NM_003666.3 | NP_003657.1 | 4 | 1141 | |||
XM_005245561.4 | XP_005245618.1 | 4 | 1048 | |||
XM_011510092.2 | XP_011508394.1 | 4 | 1044 | |||
GenBank mRNA | AK223533.1 | - | 4 | 1003 | ||
AK291725.1 | - | 4 | 808 | |||
AK307151.1 | - | 5 | 649 | |||
BC020716.1 | - | 4 | 790 | 4723421 | ||
U79751.1 | - | 4 | 1030 |