Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE8B SIRNA Assays ›
Gene Symbol:
PDE8B
Gene Name:
phosphodiesterase 8B
Gene Aliases:
ADSD, PPNAD3
Chromosome Location:
Chr.5: 77180252 - 77428256 on Build GRCh38
UniGene ID:
Hs.584830
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139349
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE8B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001029851.2
NP_001025022.1
6
781
NM_001029852.2
NP_001025023.1
6
781
NM_001029853.2
NP_001025024.1
5
721
NM_001029854.2
NP_001025025.1
6
781
NM_003719.3
NP_003710.1
6
781
XM_005248621.4
XP_005248678.1
6
1007
XM_005248623.4
XP_005248680.1
6
632
XM_005248624.4
XP_005248681.1
6
576
XM_006714725.3
XP_006714788.1
7
1074
XM_006714726.3
XP_006714789.1
7
1071
XM_011543699.2
XP_011542001.1
7
1076
XM_011543700.2
XP_011542002.1
7
1075
XM_011543701.2
XP_011542003.1
7
587
XM_011543702.2
XP_011542004.1
6
418
XM_011543703.2
XP_011542005.1
6
436
XM_011543704.2
XP_011542006.1
4
239
XM_017010003.1
XP_016865492.1
6
1009
XM_017010004.1
XP_016865493.1
6
415
XM_017010005.1
XP_016865494.1
6
1008
XM_017010006.1
XP_016865495.1
7
854
XM_017010007.1
XP_016865496.1
6
622
XM_017010008.1
XP_016865497.1
6
622
GenBank mRNA
AB085824.1
-
6
745
AB085825.1
-
6
742
AB085826.1
-
6
742
AB085827.1
-
5
682
AF079529.2
-
6
781
AL831924.1
-
6
611
AY129948.1
-
6
781
AY129949.1
-
6
781
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_199268, XM_008760669, XM_006231811, XM_008760670, AB092697.This siRNA also targets the following transcripts in Mouse: NM_001170669, NM_172263, EF644173, AK043680, AK045991, and others.
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
cAMP catabolic process
signal transduction
cyclic nucleotide metabolic process
negative regulation of insulin secretion
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline