Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003726.3 | NP_003717.3 | 10, 11 | 994 | ||
XM_017025257.1 | XP_016880746.1 | 9, 10 | 916 | |||
XM_017025258.1 | XP_016880747.1 | 7, 8 | 1123 | |||
XM_017025259.1 | XP_016880748.1 | 11, 12 | 1131 | |||
XM_017025260.1 | XP_016880749.1 | 7, 8 | 696 | |||
GenBank mRNA | AK290818.1 | - | 10, 11 | 981 | ||
Y11215.1 | - | 10, 11 | 952 |