Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BAG6 SIRNA Assays ›
Gene Symbol:
BAG6
Gene Name:
BCL2 associated athanogene 6
Gene Aliases:
BAG-6, BAT3, D6S52E, G3
Chromosome Location:
Chr.6: 31639028 - 31660900 on Build GRCh38
UniGene ID:
Hs.440900
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139431
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BAG6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098534.1
NP_001092004.1
6
701
NM_001199697.1
NP_001186626.1
6
831
NM_001199698.1
NP_001186627.1
6
831
NM_004639.3
NP_004630.3
6
855
NM_080702.2
NP_542433.1
6
831
NM_080703.2
NP_542434.1
6
855
XM_011514892.2
XP_011513194.1
6
1814
XM_017011275.1
XP_016866764.1
6
1814
XM_017011276.1
XP_016866765.1
6
804
XM_017011277.1
XP_016866766.1
6
664
XM_017011278.1
XP_016866767.1
6
739
XM_017011279.1
XP_016866768.1
6
1134
XM_017011280.1
XP_016866769.1
6
706
XM_017011281.1
XP_016866770.1
6
868
XM_017011282.1
XP_016866771.1
6
660
XM_017011283.1
XP_016866772.1
6
1814
XM_017011284.1
XP_016866773.1
6
1814
XM_017011285.1
XP_016866774.1
5
604
XM_017011286.1
XP_016866775.1
5
623
XM_017011287.1
XP_016866776.1
6
1814
XM_017011288.1
XP_016866777.1
5
604
XM_017011289.1
XP_016866778.1
6
1814
XM_017011290.1
XP_016866779.1
6
1814
XM_017011291.1
XP_016866780.1
6
1814
XM_017011292.1
XP_016866781.1
6
1814
XM_017011293.1
XP_016866782.1
5
604
XM_017011294.1
XP_016866783.1
6
1814
XM_017011295.1
XP_016866784.1
6
1814
XM_017011296.1
XP_016866785.1
6
1814
XM_017011297.1
XP_016866786.1
5
604
XM_017011298.1
XP_016866787.1
6
1814
GenBank mRNA
AK302695.1
-
6
782
AK304879.1
-
6
782
BC003133.1
-
6
749
2957896
BX384328.2
-
6
834
BX647244.1
-
6
709
M33519.1
-
6
790
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
kidney development
immune system process
ubiquitin-dependent protein catabolic process
transport
synaptonemal complex assembly
spermatogenesis
brain development
embryo development
covalent chromatin modification
internal peptidyl-lysine acetylation
cell differentiation
lung development
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of cell proliferation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of apoptotic process
negative regulation of proteolysis
protein stabilization
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
tail-anchored membrane protein insertion into ER membrane
positive regulation of ERAD pathway
maintenance of unfolded protein involved in ERAD pathway
protein localization to cytosolic proteasome complex involved in ERAD pathway
protein binding
Hsp70 protein binding
polyubiquitin binding
ubiquitin protein ligase binding
ribosome binding
misfolded protein binding
proteasome binding
ubiquitin-specific protease binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0