Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLLT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195626.1 | NP_001182555.1 | 9 | 814 | ||
NM_001324297.1 | NP_001311226.1 | 12 | 1647 | |||
NM_004641.3 | NP_004632.1 | 10 | 1163 | |||
NR_136736.1 | 11 | 1341 | ||||
GenBank mRNA | AB209755.1 | - | 11 | 1047 | ||
AY598745.1 | - | 9 | 814 | |||
BC129946.1 | - | 10 | 857 | |||
BX648210.1 | - | 3 | 190 | |||
U13948.1 | - | 10 | 997 |
Gene Symbol: PICALM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF051633.1 | - | 276 |