Gene Symbol: USP9Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004654.3 | NP_004645.2 | 38, 39 | 7391 | ||
XM_017030078.1 | XP_016885567.1 | 37, 38 | 6546 | |||
GenBank mRNA | AF000986.2 | - | 38, 39 | 7391 | ||
Y13618.1 | - | 36, 37 | 6503 | |||
Y13619.1 | - | 36, 37 | 6518 |