Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025252.2 | NP_001020423.1 | 6 | 1541 | ||
NM_001025253.2 | NP_001020424.1 | 8 | 1328 | |||
NM_001287140.1 | NP_001274069.1 | 8 | 1610 | |||
NM_001287142.1 | NP_001274071.1 | 7 | 1583 | |||
NM_001287143.1 | NP_001274072.1 | 7 | 1568 | |||
NM_001287144.1 | NP_001274073.1 | 6 | 1252 | |||
NM_005079.3 | NP_005070.1 | 6 | 1259 | |||
NR_105033.1 | 9 | 1680 | ||||
NR_105034.1 | 5 | 1143 | ||||
NR_105035.1 | 7 | 1329 | ||||
NR_105036.1 | 8 | 1273 | ||||
NR_105037.1 | 7 | 1274 | ||||
GenBank mRNA | AF202897.1 | - | 6 | 1485 | ||
AK057075.1 | - | 8 | 1329 | |||
AK295468.1 | - | 8 | 1195 | |||
AK296615.1 | - | 6 | 1263 | |||
AK308983.1 | - | 8 | 1185 | |||
BC018117.1 | - | 6 | 1130 | 3916870 | ||
BX640835.1 | - | 6 | 1325 | |||
GQ499324.1 | - | 8 | 1595 | |||
GQ499325.1 | - | 7 | 1568 | |||
GQ499326.1 | - | 7 | 1553 | |||
GQ499327.1 | - | 6 | 1526 | |||
GQ499328.1 | - | 9 | 1665 | |||
S82081.1 | - | 6 | 1136 | |||
U18914.1 | - | 6 | 1134 |