Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035247.2 | NP_001030324.2 | 5, 6 | 556 | ||
NM_005659.6 | NP_005650.2 | 5, 6 | 556 | |||
XM_011530361.2 | XP_011528663.1 | 5, 6 | 682 | |||
GenBank mRNA | AF141201.1 | - | 4, 5 | 426 | ||
AJ239058.1 | - | 4, 5 | 426 | |||
AK225877.1 | - | 5, 6 | 513 | |||
AK304734.1 | - | 5, 6 | 526 | |||
AK308786.1 | - | 5, 6 | 500 | |||
AK315998.1 | - | 5, 6 | 507 | |||
AY101594.1 | - | 5, 6 | 483 | |||
BC001049.2 | - | 5, 6 | 503 | 3507963 | ||
BC005087.1 | - | 5, 6 | 504 | 3830774 | ||
BM761958.1 | - | 4, 5 | 495 | |||
BX648483.1 | - | 5, 6 | 488 | |||
CR456607.1 | - | 5, 6 | 440 | |||
U64444.1 | - | 612 |