Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005664.3 | NP_005655.1 | 1 | 1329 | ||
GenBank mRNA | AK309383.1 | - | 1 | 974 | ||
BC044639.1 | - | 1 | 952 | 4140028 |