Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC1 SIRNA Assays ›
Gene Symbol:
TACC1
Gene Name:
transforming acidic coiled-coil containing protein 1
Gene Aliases:
Ga55
Chromosome Location:
Chr.8: 38728186 - 38853028 on Build GRCh38
UniGene ID:
Hs.279245
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139592
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146216.2
NP_001139688.1
3
1624
NM_006283.2
NP_006274.2
3
1678
XM_005273625.4
XP_005273682.1
4
1792
XM_005273628.1
XP_005273685.1
3
1299
XM_005273629.1
XP_005273686.1
4
1312
XM_005273630.1
XP_005273687.1
3
1320
XM_011544631.1
XP_011542933.1
3
1678
XM_011544632.2
XP_011542934.1
5
1853
XM_011544633.1
XP_011542935.1
3
1678
XM_011544634.1
XP_011542936.1
3
1678
XM_011544635.1
XP_011542937.1
3
1382
XM_017013766.1
XP_016869255.1
3
1299
XM_017013767.1
XP_016869256.1
4
1312
XM_017013768.1
XP_016869257.1
3
1299
XM_017013769.1
XP_016869258.1
4
1312
XM_017013770.1
XP_016869259.1
2
1212
XM_017013771.1
XP_016869260.1
2
1484
XM_017013772.1
XP_016869261.1
2
1212
XM_017013773.1
XP_016869262.1
2
1479
XM_017013774.1
XP_016869263.1
2
1164
XM_017013775.1
XP_016869264.1
3
1320
XM_017013776.1
XP_016869265.1
4
1312
XM_017013777.1
XP_016869266.1
2
1476
XM_017013778.1
XP_016869267.1
2
1175
GenBank mRNA
AB029026.1
-
1
212
AF049910.1
-
3
1619
AK294931.1
-
3
1279
AK295071.1
-
3
1226
AK295841.1
-
2
1215
AK303596.1
-
2
1110
AK304507.1
-
5
1511
AK314620.1
-
3
1678
AY072876.2
-
3
1230
BC041391.1
-
5
1875
5276767
BX647696.1
-
3
1626
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell cycle
cell proliferation
cerebral cortex development
cell division
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-8tvtn:80/10.239.165.222:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0