Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006767.3 | NP_006758.2 | 6 | 653 | ||
GenBank mRNA | AK296347.1 | - | 5 | 534 | ||
AK310747.1 | - | 4 | 256 | |||
AK313502.1 | - | 1 | 55 | |||
BC026214.1 | - | 6 | 555 | 4399716 | ||
CT841521.1 | - | 6 | 569 | |||
D38496.1 | - | 6 | 547 |