Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012465.3 | NP_036597.1 | 20 | 3045 | ||
GenBank mRNA | AB023149.2 | - | 20 | 2995 | ||
AF059516.1 | - | 20 | 3168 | |||
BC112341.1 | - | 20 | 2928 | 40038720 | ||
BC112366.1 | - | 20 | 2928 | 40038726 | ||
BC113577.1 | - | 20 | 2861 | 8322629 |