Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003218.3 | NP_003209.2 | 8 | 1077 | ||
NM_017489.2 | NP_059523.2 | 9 | 1137 | |||
XM_005251291.3 | XP_005251348.1 | 10 | 1225 | |||
XM_005251292.3 | XP_005251349.1 | 9 | 1165 | |||
XM_011517582.2 | XP_011515884.1 | 11 | 1292 | |||
XM_017013792.1 | XP_016869281.1 | 9 | 975 | |||
GenBank mRNA | AF003001.1 | - | 8 | 1067 | ||
AK123410.1 | - | 2 | 1348 | |||
AK292529.1 | - | 9 | 1135 | |||
BC029378.1 | - | 8 | 1060 | 4724068 | ||
U40705.1 | - | 9 | 1127 | |||
U74382.1 | - | 8 | 1052 | |||
X93511.1 | - | 2 | 128 |