Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145128.2 | NP_001138600.2 | 10 | 991 | ||
NM_145025.4 | NP_659462.1 | 10 | 991 | |||
XM_006715376.3 | XP_006715439.2 | 11 | 1856 | |||
XM_011535550.2 | XP_011533852.1 | 11 | 1856 | |||
XM_011535552.2 | XP_011533854.1 | 10 | 1032 | |||
XM_011535553.2 | XP_011533855.1 | 10 | 1639 | |||
XM_011535554.2 | XP_011533856.1 | 11 | 1856 | |||
XM_011535555.2 | XP_011533857.1 | 11 | 1856 | |||
XM_011535556.2 | XP_011533858.1 | 11 | 1856 | |||
XM_011535559.2 | XP_011533861.1 | 11 | 1856 | |||
XM_017010384.1 | XP_016865873.1 | 11 | 1856 | |||
XM_017010385.1 | XP_016865874.1 | 10 | 1024 | |||
XM_017010386.1 | XP_016865875.1 | 11 | 1856 | |||
XM_017010387.1 | XP_016865876.1 | 5 | 577 | |||
XM_017010388.1 | XP_016865877.1 | 11 | 1856 | |||
GenBank mRNA | AK055538.1 | - | 10 | 968 | ||
AK315561.1 | - | 10 | 962 | |||
AL832162.1 | - | 10 | 970 | |||
BC022031.2 | - | 10 | 996 |