Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199237.1 | NP_001186166.1 | 3 | 996 | ||
NM_001199238.1 | NP_001186167.1 | 3 | 865 | |||
NM_001199239.1 | NP_001186168.1 | 3 | 862 | |||
NM_001199240.1 | NP_001186169.1 | 3 | 648 | |||
NM_033177.3 | NP_149417.1 | 2 | 972 | |||
XM_005249403.3 | XP_005249460.1 | 3 | 999 | |||
XM_006715204.1 | XP_006715267.1 | 3 | 677 | |||
XM_011514909.1 | XP_011513211.1 | 3 | 717 | |||
XM_011514910.1 | XP_011513212.1 | 3 | 714 | |||
GenBank mRNA | AJ420471.1 | - | 2 | 564 | ||
AK123707.1 | - | 3 | 996 | |||
AK314056.1 | - | 3 | 865 | |||
BC008783.1 | - | 2 | 962 | 3629572 | ||
BC018875.2 | - | 2 | 956 |