Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199237.1 | NP_001186166.1 | 4 | 1576 | ||
NM_001199238.1 | NP_001186167.1 | 4 | 1445 | |||
NM_001199239.1 | NP_001186168.1 | 4 | 1442 | |||
NM_001199240.1 | NP_001186169.1 | 4 | 1228 | |||
NM_033177.3 | NP_149417.1 | 3 | 1552 | |||
XM_005249403.3 | XP_005249460.1 | 4 | 1579 | |||
XM_006715204.1 | XP_006715267.1 | 4 | 1257 | |||
XM_011514909.1 | XP_011513211.1 | 4 | 1297 | |||
XM_011514910.1 | XP_011513212.1 | 4 | 1294 | |||
GenBank mRNA | AJ420471.1 | - | 3 | 1144 | ||
AK123707.1 | - | 4 | 1576 | |||
AK314056.1 | - | 4 | 1445 | |||
BC008783.1 | - | 3 | 1542 | 3629572 | ||
BC018875.2 | - | 3 | 1536 |