Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023560.3 | NP_001018854.2 | 4 | 1016 | ||
NM_001111039.2 | NP_001104509.1 | 4 | 1013 | |||
NM_001287421.1 | NP_001274350.1 | 4 | 502 | |||
NM_001287422.1 | NP_001274351.1 | 2 | 412 | |||
NM_152736.5 | NP_689949.3 | 4 | 611 | |||
XM_011514862.2 | XP_011513164.1 | 4 | 1215 | |||
XM_011514864.1 | XP_011513166.1 | 5 | 792 | |||
XM_011514866.1 | XP_011513168.1 | 4 | 564 | |||
XM_011514867.1 | XP_011513169.1 | 4 | 928 | |||
XM_011514869.1 | XP_011513171.1 | 5 | 856 | |||
XM_017011264.1 | XP_016866753.1 | 4 | 1213 | |||
XM_017011265.1 | XP_016866754.1 | 4 | 947 | |||
GenBank mRNA | AK095948.1 | - | 4 | 563 | ||
AK293524.1 | - | 4 | 736 | |||
AK295393.1 | - | 3 | 1166 | |||
AK297613.1 | - | 2 | 273 | |||
AK301081.1 | - | 4 | 907 | |||
AK315645.1 | - | 4 | 475 | |||
AL526328.3 | - | 4 | 868 | |||
AL832741.1 | - | 3 | 1204 | |||
BC013951.2 | - | 4 | 467 | 2963683 | ||
BC013962.2 | - | 4 | 467 | 2963683 | ||
BX537535.1 | - | 4 | 884 | |||
DA167287.1 | - | 4 | 500 | |||
DA341149.1 | - | 4 | 500 | |||
DA528922.1 | - | 2 | 273 | |||
DB262147.1 | - | 4 | 478 | |||
Z11773.1 | - | 3 | 533 |