Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143887.1 | NP_001137359.1 | 2 | 232 | ||
NM_004236.3 | NP_004227.1 | 2 | 232 | |||
GenBank mRNA | AB209799.1 | - | 2 | 158 | ||
AF084260.1 | - | 2 | 168 | |||
AF100762.1 | - | 2 | 158 | |||
AF120268.1 | - | 2 | 144 | |||
AF212227.1 | - | 2 | 158 | |||
AK222590.1 | - | 2 | 156 | |||
AK312643.1 | - | 2 | 168 | |||
BC012629.1 | - | 2 | 169 | 4245220 | ||
CR542063.1 | - | 2 | 153 | |||
DB059510.1 | - | 2 | 236 | |||
L40388.1 | - | 2 | 144 |