Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNDC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144825.1 | NP_001138297.1 | 6 | 887 | ||
NM_001144826.1 | NP_001138298.1 | 6 | 872 | |||
NM_006695.4 | NP_006686.1 | 6 | 887 | |||
XM_005256964.3 | XP_005257021.1 | 6 | 820 | |||
XM_017024035.1 | XP_016879524.1 | 6 | 887 | |||
XM_017024036.1 | XP_016879525.1 | 6 | 808 | |||
XM_017024037.1 | XP_016879526.1 | 6 | 820 | |||
XM_017024038.1 | XP_016879527.1 | 6 | 887 | |||
XM_017024039.1 | XP_016879528.1 | 6 | 887 | |||
XM_017024040.1 | XP_016879529.1 | 6 | 808 | |||
GenBank mRNA | AB209802.1 | - | 6 | 871 | ||
AF055026.1 | - | 6 | 651 | |||
AK123330.1 | - | 5 | 2532 | |||
AK226045.1 | - | 6 | 848 | |||
AK313669.1 | - | 6 | 808 | |||
AL133657.1 | - | 6 | 1033 | |||
BC006194.2 | - | 6 | 850 | 2823366 | ||
BX537443.1 | - | 6 | 769 | |||
CR456748.1 | - | 6 | 613 | |||
U93871.1 | - | 6 | 598 |