Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBGCP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286414.2 | NP_001273343.1 | 18 | 2443 | ||
NM_014444.4 | NP_055259.2 | 18 | 2440 | |||
XM_011521454.2 | XP_011519756.1 | 17 | 2326 | |||
XM_011521455.2 | XP_011519757.1 | 18 | 2341 | |||
XM_017022078.1 | XP_016877567.1 | 17 | 2323 | |||
GenBank mRNA | AK001639.1 | - | 14 | 1754 | ||
AK026551.1 | - | 18 | 2430 | |||
AK027703.1 | - | 18 | 2401 | |||
BC012801.2 | - | 18 | 2384 | 2821891 |