Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286479.1 | NP_001273408.1 | 3 | 257 | ||
NM_001286480.1 | NP_001273409.1 | 4 | 379 | |||
NM_014783.4 | NP_055598.1 | 3 | 998 | |||
NM_199357.1 | NP_955389.1 | 3 | 998 | |||
GenBank mRNA | AK303025.1 | - | 4 | 298 | ||
AK316437.1 | - | 3 | 257 | |||
BC039563.2 | - | 3 | 853 | 3913511 | ||
BC050545.1 | - | 3 | 953 | |||
BC063444.1 | - | 3 | 949 | |||
D87717.2 | - | 3 | 998 |
Gene Symbol: ARHGAP11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039841.1 | NP_001034930.1 | 3 | 421 | ||
GenBank mRNA | AK310319.1 | - | 4 | 306 | ||
BC105788.1 | - | 3 | 421 | 4072723 |