Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HDM2 SIRNA Assays ›
Gene Symbol:
R3HDM2
Gene Name:
R3H domain containing 2
Gene Aliases:
CAG6, PR01365
Chromosome Location:
Chr.12: 57253764 - 57430999 on Build GRCh38
UniGene ID:
Hs.443673
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
140206
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HDM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014925.3
NP_055740.3
22
2960
XM_005268725.3
XP_005268782.1
24
3310
XM_011538031.1
XP_011536333.1
25
3267
XM_011538033.1
XP_011536335.1
26
3252
XM_011538035.1
XP_011536337.1
25
3225
XM_011538036.1
XP_011536338.1
25
3219
XM_011538037.1
XP_011536339.1
24
3201
XM_011538038.1
XP_011536340.1
24
3171
XM_011538039.1
XP_011536341.1
24
3165
XM_011538040.1
XP_011536342.1
25
3150
XM_011538042.2
XP_011536344.1
27
3417
XM_017018998.1
XP_016874487.1
24
3243
XM_017018999.1
XP_016874488.1
27
3458
XM_017019000.1
XP_016874489.1
25
3156
XM_017019001.1
XP_016874490.1
26
3531
XM_017019002.1
XP_016874491.1
24
3126
XM_017019003.1
XP_016874492.1
24
3102
XM_017019004.1
XP_016874493.1
24
3096
XM_017019005.1
XP_016874494.1
26
3307
XM_017019006.1
XP_016874495.1
25
3200
XM_017019007.1
XP_016874496.1
25
3235
XM_017019008.1
XP_016874497.1
23
3069
XM_017019009.1
XP_016874498.1
24
3054
XM_017019010.1
XP_016874499.1
24
3197
XM_017019011.1
XP_016874500.1
27
3351
XM_017019012.1
XP_016874501.1
23
3036
XM_017019013.1
XP_016874502.1
24
3170
XM_017019014.1
XP_016874503.1
24
3205
XM_017019015.1
XP_016874504.1
24
3416
XM_017019016.1
XP_016874505.1
25
3366
XM_017019017.1
XP_016874506.1
24
3128
XM_017019018.1
XP_016874507.1
24
3094
XM_017019019.1
XP_016874508.1
25
3190
XM_017019020.1
XP_016874509.1
22
2940
XM_017019021.1
XP_016874510.1
24
3164
XM_017019022.1
XP_016874511.1
23
3074
XM_017019023.1
XP_016874512.1
23
3085
XM_017019024.1
XP_016874513.1
23
3074
XM_017019025.1
XP_016874514.1
23
3039
XM_017019026.1
XP_016874515.1
24
3079
XM_017019027.1
XP_016874516.1
23
3210
XM_017019028.1
XP_016874517.1
22
2972
XM_017019029.1
XP_016874518.1
22
3058
XM_017019030.1
XP_016874519.1
15
2339
XM_017019031.1
XP_016874520.1
18
2623
XM_017019032.1
XP_016874521.1
14
2191
XM_017019033.1
XP_016874522.1
16
2342
GenBank mRNA
AB023219.1
-
24
3317
AF113695.1
-
1
164
AX775761.1
-
24
3317
AX775763.1
-
24
3359
BC008143.1
-
1
173
BC041857.1
-
14
2772
5270511
BC104995.1
-
12
1967
8143998
BC112226.1
-
12
1967
8327694
BC143605.1
-
13
2069
9052118
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0