Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015134.3 | NP_055949.2 | 20 | 2824 | ||
NM_201274.3 | NP_958431.2 | 20 | 2824 | |||
XM_005256563.4 | XP_005256620.1 | 21 | 6936 | |||
XM_005256564.4 | XP_005256621.3 | 19 | 3015 | |||
XM_011523761.2 | XP_011522063.1 | 21 | 7307 | |||
XM_011523762.2 | XP_011522064.1 | 21 | 7290 | |||
XM_011523763.2 | XP_011522065.1 | 20 | 7176 | |||
XM_011523764.2 | XP_011522066.1 | 20 | 7170 | |||
XM_011523765.2 | XP_011522067.1 | 20 | 7170 | |||
XM_011523766.2 | XP_011522068.2 | 20 | 3483 | |||
XM_011523767.2 | XP_011522069.2 | 20 | 3483 | |||
XM_017024393.1 | XP_016879882.1 | 19 | 3363 | |||
GenBank mRNA | AB020671.2 | - | 6 | 3828 | ||
AB098507.1 | - | 20 | 2788 | |||
AB189741.1 | - | 20 | 2729 | |||
AB362572.1 | - | 3 | 204 | |||
AB362573.1 | - | 3 | 204 | |||
AJ844625.1 | - | 3 | 198 | |||
AK025229.1 | - | 10 | 1489 | |||
AK025604.1 | - | 6 | 2931 | |||
AK092287.1 | - | 15 | 2202 | |||
AL834513.1 | - | 19 | 2621 | |||
AY296247.1 | - | 20 | 2732 | |||
BC009982.2 | - | 15 | 2147 | |||
BC012426.1 | - | 2 | 73 | |||
BC021058.1 | - | 7 | 732 | |||
BC021070.1 | - | 7 | 732 | |||
BC075847.1 | - | 15 | 2158 | |||
BC105987.1 | - | 12 | 1608 | |||
BC136479.1 | - | 20 | 2812 | 9020468 | ||
D23673.1 | - | 6 | 701 |