Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESYT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184796.1 | NP_001171725.1 | 27 | 3117 | ||
NM_015292.2 | NP_056107.1 | 27 | 3087 | |||
GenBank mRNA | AB018290.1 | - | 27 | 2874 | ||
AK025463.1 | - | 23 | 2292 | |||
AK025878.1 | - | 8 | 779 | |||
AK074368.1 | - | 16 | 1615 | |||
AK128730.1 | - | 26 | 2891 | |||
AK225035.1 | - | 27 | 3052 | |||
AK290337.1 | - | 27 | 3022 | |||
AL050134.1 | - | 19 | 1835 | |||
BC004998.1 | - | 27 | 3019 | 2958094 | ||
DQ993200.1 | - | 27 | 2969 |