Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015336.2 | NP_056151.2 | 9 | 1137 | ||
XM_005268750.4 | XP_005268807.1 | 9 | 936 | |||
XM_011538083.1 | XP_011536385.1 | 9 | 1001 | |||
XM_017019091.1 | XP_016874580.1 | 10 | 1087 | |||
XM_017019092.1 | XP_016874581.1 | 9 | 950 | |||
XM_017019093.1 | XP_016874582.1 | 9 | 899 | |||
XM_017019094.1 | XP_016874583.1 | 8 | 817 | |||
XM_017019095.1 | XP_016874584.1 | 5 | 529 | |||
GenBank mRNA | AB023163.1 | - | 9 | 1081 | ||
AB024494.1 | - | 9 | 1627 | |||
AB097013.1 | - | 9 | 957 | |||
AB097035.1 | - | 9 | 957 | |||
AK292866.1 | - | 9 | 953 | |||
AK299089.1 | - | 9 | 959 | |||
BC050324.1 | - | 9 | 1096 | 5266097 |