Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TBC1D10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015527.3 | NP_056342.3 | 4 | 1268 | ||
XM_011545790.2 | XP_011544092.1 | 4 | 1325 | |||
GenBank mRNA | AB449895.1 | - | 4 | 785 | ||
AF370370.1 | - | 4 | 413 | |||
AK023192.1 | - | 4 | 647 | |||
BC063112.1 | - | 4 | 692 | |||
BC072453.1 | - | 4 | 580 | 6184429 | ||
BC093814.1 | - | 4 | 496 | 7939659 | ||
BC093816.1 | - | 4 | 496 | 7939661 | ||
BC098419.1 | - | 4 | 832 |