Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305077.1 | NP_001292006.1 | 7 | 1933 | ||
NM_001305078.1 | NP_001292007.1 | 7 | 1932 | |||
NM_001305079.1 | NP_001292008.1 | 7 | 1859 | |||
NM_001305080.1 | NP_001292009.1 | 7 | 1921 | |||
NM_001305081.1 | NP_001292010.1 | 7 | 1930 | |||
NM_001305082.1 | NP_001292011.1 | 7 | 2081 | |||
NM_001305083.1 | NP_001292012.1 | 7 | 1918 | |||
NM_001305084.1 | NP_001292013.1 | 6 | 1817 | |||
NM_001305085.1 | NP_001292014.1 | 6 | 1814 | |||
NM_001305086.1 | NP_001292015.1 | 6 | 1838 | |||
NM_001305087.1 | NP_001292016.1 | 6 | 1837 | |||
NM_001305088.1 | NP_001292017.1 | 6 | 1770 | |||
NM_015654.4 | NP_056469.2 | 7 | 1936 | |||
NR_130953.1 | 8 | 1940 | ||||
NR_130954.1 | 7 | 1818 | ||||
XM_005257212.4 | XP_005257269.1 | 6 | 1775 | |||
XM_006721813.3 | XP_006721876.1 | 6 | 2223 | |||
XM_017024437.1 | XP_016879926.1 | 7 | 1862 | |||
XM_017024439.1 | XP_016879928.1 | 7 | 1826 | |||
XM_017024441.1 | XP_016879930.1 | 5 | 2116 | |||
GenBank mRNA | AK123115.1 | - | 7 | 2003 | ||
AK225602.1 | - | 8 | 1721 | |||
AY632082.1 | - | 6 | 1772 | |||
BC004195.2 | - | 7 | 1842 | 3528894 | ||
BC004225.2 | - | 7 | 1831 | 3538186 |