Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303447.1 | NP_001290376.1 | 9 | 1674 | ||
NM_001324022.1 | NP_001310951.1 | 7 | 1463 | |||
NM_001324024.1 | NP_001310953.1 | 7 | 1480 | |||
NM_001324025.1 | NP_001310954.1 | 6 | 1350 | |||
NM_015914.6 | NP_056998.4 | 8 | 1593 | |||
XM_011522515.2 | XP_011520817.1 | 9 | 1670 | |||
XM_011522516.2 | XP_011520818.1 | 8 | 1430 | |||
XM_017023268.1 | XP_016878757.1 | 9 | 1667 | |||
GenBank mRNA | AK027464.1 | - | 1 | 176 | ||
AK027646.1 | - | 8 | 1507 | |||
AK074534.1 | - | 1 | 498 | |||
AK297094.1 | - | 8 | 1061 | |||
BC002856.1 | - | 9 | 1643 | 3635977 | ||
BX648407.1 | - | 9 | 1300 |