Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205028.1 | NP_001191957.1 | 3 | 377 | ||
NM_001205029.1 | NP_001191958.1 | 3 | 866 | |||
NM_017440.4 | NP_059136.2 | 3 | 377 | |||
NM_020128.2 | NP_064513.1 | 3 | 377 | |||
XM_005269026.1 | XP_005269083.1 | 3 | 302 | |||
XM_011538569.1 | XP_011536871.1 | 3 | 302 | |||
XM_017019646.1 | XP_016875135.1 | 3 | 298 | |||
XM_017019647.1 | XP_016875136.1 | 3 | 302 | |||
XM_017019648.1 | XP_016875137.1 | 3 | 809 | |||
XM_017019649.1 | XP_016875138.1 | 3 | 298 | |||
XM_017019650.1 | XP_016875139.1 | 3 | 809 | |||
GenBank mRNA | AF267851.1 | - | 3 | 299 | ||
AK297311.1 | - | 3 | 282 | |||
AK314461.1 | - | 3 | 356 | |||
BC028355.2 | - | 3 | 328 | 4827410 | ||
BM787113.1 | - | 3 | 377 | |||
CD704611.1 | - | 3 | 380 |